1-{4-[(3,4-Dichlorobenzyl)oxy]phenyl}-1-ethanone ≥95%

Supplier: Matrix Scientific

047608-500MG 047608-1G
101880-766EA 116.92 USD
101880-766 101880-768
1-{4-[(3,4-Dichlorobenzyl)oxy]phenyl}-1-ethanone ≥95%
1-{4-[(3,4-Dichlorobenzyl)oxy]phenyl}-1-ethanone

Hazard: Irritant

Formula: C₁₅H₁₂Cl₂O₂
Melting Pt: 83…85 °C
MDL Number: MFCD00173669
CAS Number: 170916-55-9

Order Now

Specification Test Results

Melting point 83-85°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR