Methyl-3-(4-hydroxyphenyl)acrylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-(4-hydroxyphenyl)acrylate, Methyl 4-hydroxycinnamate

047607-1G 047607-5G 047607-25G
101880-760EA 775.38 USD
101880-760 101880-762 101880-764
Methyl-3-(4-hydroxyphenyl)acrylate ≥95%
Methyl-3-(4-hydroxyphenyl)acrylate

Hazard: Irritant

Formula: C₁₀H₁₀O₃
MW: 178.1846 g/mol
Melting Pt: 144…145 °C
MDL Number: MFCD00157167
CAS Number: 3943-97-3

Order Now

Specification Test Results

Melting point 144-145°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR