3-Phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%

Supplier: Matrix Scientific

047602-500MG 047602-1G 047602-5G
101880-740EA 101.54 USD
101880-740 101880-742 101880-744
3-Phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
3-Phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone

Hazard: Irritant

Formula: C₁₄H₁₀N₂OS
MW: 254.31 g/mol
Melting Pt: 313…314 °C
MDL Number: MFCD00023895
CAS Number: 18741-24-7

Order Now

Specification Test Results

Melting point 313-314°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR