1-(4-Iodophenyl)-1H-pyrrole-2,5-dione ≥95%

Supplier: Matrix Scientific

047599-500MG 047599-1G
101880-728EA 169.23 USD
101880-728 101880-730
1-(4-Iodophenyl)-1H-pyrrole-2,5-dione ≥95%
1-(4-Iodophenyl)-1H-pyrrole-2,5-dione

Hazard: Irritant

Formula: C₁₀H₆INO₂
Melting Pt: 160 °C
MDL Number: MFCD00022575
CAS Number: 65833-01-4

Order Now

Specification Test Results

Melting point 160°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR