2-(3-Chloro-4-fluoroanilino)acetohydrazide ≥95%

Supplier: Matrix Scientific

047593-500MG 047593-1G 047593-5G
101880-704EA 176.92 USD
101880-704 101880-706 101880-708
2-(3-Chloro-4-fluoroanilino)acetohydrazide ≥95%
2-(3-Chloro-4-fluoroanilino)acetohydrazide

Hazard: Irritant

Formula: C₈H₉ClFN₃O
Melting Pt: 99…100 °C
MDL Number: MFCD03788387
CAS Number: 2370-44-7

Order Now

Specification Test Results

Melting point 99-100°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR