3-[4-(Methylsulfanyl)phenyl]acrylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(4-(Methylthio)phenyl)acrylic acid, 4-(Methylthio)cinnamic acid

047588-500MG 047588-1G 047588-5G
101880-682EA 153.85 USD
101880-682 101880-684 101880-686
3-[4-(Methylsulfanyl)phenyl]acrylic acid ≥95%
3-[4-(Methylsulfanyl)phenyl]acrylic acid

Hazard: Irritant

Formula: C₁₀H₁₀O₂S
MW: 194.25 g/mol
Melting Pt: 174…175 °C
MDL Number: MFCD00266583
CAS Number: 102016-58-0

Order Now

Specification Test Results

Melting point 174-175°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR