4-(Trifluoromethoxy)cinnamic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethoxy)cinnamic Acid

047587-500MG 047587-1G 047587-5G
101880-676EA 181.54 USD
101880-676 101880-678 101880-680
4-(Trifluoromethoxy)cinnamic acid ≥95%
4-(Trifluoromethoxy)cinnamic acid

Hazard: Irritant

Formula: C₁₀H₇F₃O₃
MW: 232.1600 g/mol
Melting Pt: 185-186°C
MDL Number: MFCD00066338
CAS Number: 783-13-1

Order Now

Specification Test Results

Melting point 185-186°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR