3-(4-Methoxy-1-naphthyl)acrylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(4-Methoxynaphth-1-yl)-2-propenoic acid

047584-500MG 047584-1G 047584-5G
101880-660EA 181.54 USD
101880-660 101880-662 101880-664
3-(4-Methoxy-1-naphthyl)acrylic acid ≥95%
3-(4-Methoxy-1-naphthyl)acrylic acid

Hazard: Irritant

Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Melting Pt: 225 °C
MDL Number: MFCD00156955
CAS Number: 15971-30-9

Order Now

Specification Test Results

Melting point 225°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR