2-Chloro-N-(2,4,5-trifluorophenyl)acetamide ≥95%

Supplier: Matrix Scientific

047575-500MG 047575-1G
101880-652EA 181.54 USD
101880-652 101880-654
2-Chloro-N-(2,4,5-trifluorophenyl)acetamide ≥95%
2-Chloro-N-(2,4,5-trifluorophenyl)acetamide

Hazard: Irritant

Formula: C₈H₅ClF₃NO
MDL Number: MFCD00077477
CAS Number: 885267-45-8

Order Now

Specification Test Results

Melting point 111.5-112.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR