5-[(3-Methylbenzyl)sulfanyl]-1,3,4-thiadiazol-2-ylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-(3-methylbenzylthio)-1,3,4-thiadiazole

047571-500MG 047571-1G
101880-638EA 181.54 USD
101880-638 101880-640
5-[(3-Methylbenzyl)sulfanyl]-1,3,4-thiadiazol-2-ylamine ≥95%
5-[(3-Methylbenzyl)sulfanyl]-1,3,4-thiadiazol-2-ylamine

Hazard: Irritant

Formula: C₁₀H₁₁N₃S₂
MW: 237.35 g/mol
Melting Pt: 127…129 °C
MDL Number: MFCD00448333
CAS Number: 299442-42-5

Order Now

Specification Test Results

Melting point 127-129°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR