5-(2-Propynylsulfanyl)-1,3,4-thiadiazol-2-ylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-[(propyn-3-yl)thio]-1,3,4-thiadiazole

047569-500MG 047569-1G
101880-630EA 118.46 USD
101880-630 101880-632
5-(2-Propynylsulfanyl)-1,3,4-thiadiazol-2-ylamine ≥95%
5-(2-Propynylsulfanyl)-1,3,4-thiadiazol-2-ylamine

Hazard: Irritant

Formula: C₅H₅N₃S₂
MW: 171.24 g/mol
Melting Pt: 134…136 °C
MDL Number: MFCD00175753
CAS Number: 53918-05-1

Order Now

Specification Test Results

Melting point 134-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR