3-Nitro-4-(phenylsulfanyl)benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

047566-500MG 047566-1G
101880-618EA 181.54 USD
101880-618 101880-620
3-Nitro-4-(phenylsulfanyl)benzenecarbaldehyde ≥95%
3-Nitro-4-(phenylsulfanyl)benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₉NO₃S
Melting Pt: 89…90 °C
MDL Number: MFCD02815346
CAS Number: 69054-34-8

Order Now

Specification Test Results

Melting point 89-90°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR