5-Nitro-2-(phenylsulfanyl)benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Nitro-2-(phenylthio)benzaldehyde

047565-500MG 047565-1G
101880-614EA 181.54 USD
101880-614 101880-616
5-Nitro-2-(phenylsulfanyl)benzenecarbaldehyde ≥95%
5-Nitro-2-(phenylsulfanyl)benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₉NO₃S
MW: 259.29 g/mol
Melting Pt: 106…107 °C
MDL Number: MFCD03788384
CAS Number: 52548-32-0

Order Now

Specification Test Results

Melting point 106-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR