4-[(4-Bromophenyl)sulfanyl]-3-nitrobenzenecarbaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(4-Bromophenylthio)-3-nitrobenzaldehyde
047563-500MG
047563-1G
101880-606EA
181.54
USD
101880-606
101880-608
4-[(4-Bromophenyl)sulfanyl]-3-nitrobenzenecarbaldehyde ≥95%
4-[(4-Bromophenyl)sulfanyl]-3-nitrobenzenecarbaldehyde
Hazard: Irritant
Formula:
C₁₃H₈BrNO₃S Melting Pt: 138…139 °C |
MDL Number:
MFCD00173713 CAS Number: 175278-49-6 |
Specification Test Results
Melting point | 138-139°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...