3-Nitro-4-(1-pyrrolidinyl)benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Nitro-4-(pyrrolidin-1-yl)benzaldehyde

047561-500MG 047561-1G
101880-598EA 92.31 USD
101880-598 101880-600
3-Nitro-4-(1-pyrrolidinyl)benzenecarbaldehyde ≥95%
3-Nitro-4-(1-pyrrolidinyl)benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
MW: 220.23 g/mol
Melting Pt: 107.5…108.5 °C
MDL Number: MFCD00455104
CAS Number: 284679-97-6

Order Now

Specification Test Results

Melting point 107.5-108.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR