2-Boc-aminothiazole-5-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Boc-Aminothiazole-5-carboxylic acid, 2-(tert-Butoxycarbonylamino)thiazole-5-carboxylic acid, 2-(Boc-amino)thiazole-5-carboxylic acid
047559-500MG
047559-1G
047559-5G
101880-588EA
107.69
USD
101880-588
101880-590
101880-592
2-Boc-aminothiazole-5-carboxylic acid ≥95%
2-Boc-aminothiazole-5-carboxylic acid
Hazard: Irritant
Formula:
C₉H₁₂N₂O₄S MW: 244.2676 g/mol Melting Pt: 220…222 °C (dec.) |
MDL Number:
MFCD06796614 CAS Number: 302964-02-9 |
Specification Test Results
Melting point | 220-222°C(d |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...