3-Bromo-6-chloro-2,2'-bipyridine-5-carbonitrile ≥95%

Supplier: Matrix Scientific

047557-500MG 047557-1G
101880-580EA 733.85 USD
101880-580 101880-582
3-Bromo-6-chloro-2,2'-bipyridine-5-carbonitrile ≥95%
3-Bromo-6-chloro-2,2'-bipyridine-5-carbonitrile

Hazard: Irritant

Formula: C₁₁H₅BrClN₃
Melting Pt: 130…132 °C
MDL Number: MFCD11841005
CAS Number: 1135283-47-4

Order Now

Specification Test Results

Melting point 130-132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR