tert-Butyl-4-(5-chloro-2-nitrophenyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

047552-500MG 047552-1G 047552-5G
101880-558EA 375.38 USD
101880-558 101880-560 101880-562
tert-Butyl-4-(5-chloro-2-nitrophenyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-(5-chloro-2-nitrophenyl)tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₅H₂₀ClN₃O₄
MW: 341.8 g/mol
Melting Pt: 76…78 °C
MDL Number: MFCD11553047
CAS Number: 869937-73-5

Order Now

Specification Test Results

Melting point 76-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR