N'-Cyano-3-pyridinecarboximidamide ≥95%
Supplier: Matrix Scientific
047551-500MG
047551-1G
047551-5G
101880-552EA
278.46
USD
101880-552
101880-554
101880-556
N'-Cyano-3-pyridinecarboximidamide ≥95%
N'-Cyano-3-pyridinecarboximidamide
Hazard: Irritant
Formula:
C₇H₆N₄ Melting Pt: 222…223 °C |
MDL Number:
MFCD09864976 CAS Number: 73631-23-9 |
Specification Test Results
Melting point | 222-223°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...