1-(2-Ethoxy-2-oxoethyl)-1H-1,2,3-triazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

047550-500MG 047550-1G
101880-548EA 236.92 USD
101880-548 101880-550
1-(2-Ethoxy-2-oxoethyl)-1H-1,2,3-triazole-4-carboxylic acid ≥95%
1-(2-Ethoxy-2-oxoethyl)-1H-1,2,3-triazole-4-carboxylic acid

Hazard: Irritant

Formula: C₇H₉N₃O₄
Melting Pt: 190…191 °C
MDL Number: MFCD11553045
CAS Number: 91616-28-3

Order Now

Specification Test Results

Melting point 190-191°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR