4-Hydroxy-1-(4-methoxyphenyl)-6-oxo-1,6-dihydro-3-pyridazinecarboxylic acid ≥95%

Supplier: Matrix Scientific

047549-500MG 047549-1G 047549-5G
101880-542EA 278.46 USD
101880-542 101880-544 101880-546
4-Hydroxy-1-(4-methoxyphenyl)-6-oxo-1,6-dihydro-3-pyridazinecarboxylic acid ≥95%
4-Hydroxy-1-(4-methoxyphenyl)-6-oxo-1,6-dihydro-3-pyridazinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₅
Melting Pt: 235…236 °C
MDL Number: MFCD10008589
CAS Number: 121582-69-2

Order Now

Specification Test Results

Melting point 235-236°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR