Ethyl-2-[(tert-butoxycarbonyl)amino]-1,3-thiazole-5-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-amino-1,3-thiazole-5-carboxylate, 2-BOC protected
047548-1G
047548-5G
101880-538EA
107.69
USD
101880-538
101880-540
Ethyl-2-[(tert-butoxycarbonyl)amino]-1,3-thiazole-5-carboxylate ≥95%
Ethyl-2-[(tert-butoxycarbonyl)amino]-1,3-thiazole-5-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₆N₂O₄S Melting Pt: 198…200 °C |
MDL Number:
MFCD06409216 CAS Number: 302964-01-8 |
Specification Test Results
Melting point | 198-200°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...