1-(5-Chloro-2-nitrophenyl)piperidine ≥95%

Supplier: Matrix Scientific

047545-500MG 047545-1G 047545-5G
101880-520EA 375.38 USD
101880-520 101880-522 101880-524
1-(5-Chloro-2-nitrophenyl)piperidine ≥95%
1-(5-Chloro-2-nitrophenyl)piperidine

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O₂
Melting Pt: 69…70 °C
CAS Number: 53013-43-7

Order Now

Specification Test Results

Melting point 69-70°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR