5-Bromo-2-oxo-6-(2-pyridinyl)-1,2-dihydro-3-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

047541-500MG 047541-1G 047541-5G
101880-500EA 375.38 USD
101880-500 101880-502 101880-504
5-Bromo-2-oxo-6-(2-pyridinyl)-1,2-dihydro-3-pyridinecarbonitrile ≥95%
5-Bromo-2-oxo-6-(2-pyridinyl)-1,2-dihydro-3-pyridinecarbonitrile

Hazard: Irritant

Formula: C₁₁H₆BrN₃O
MW: 276.1 g/mol
Melting Pt: 223…225 °C
MDL Number: MFCD11553040
CAS Number: 1092352-53-8

Order Now

Specification Test Results

Melting point 223-225°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR