Methyl-2-[8-iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]acetate ≥95%

Supplier: Matrix Scientific

047538-500MG 047538-1G
101880-492EA 747.69 USD
101880-492 101880-494
Methyl-2-[8-iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]acetate ≥95%
Methyl-2-[8-iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]acetate

Hazard: Irritant

Formula: C₁₁H₈F₃IN₂O₂
MW: 384.1 g/mol
Melting Pt: 126 °C
MDL Number: MFCD11553037
CAS Number: 1092352-47-0

Order Now

Specification Test Results

Melting point 126°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR