5-Amino-1-(5-chloro-2-pyridinyl)-1H-pyrazole-4-carbonitrile ≥95%

Supplier: Matrix Scientific

047536-500MG 047536-1G 047536-5G
101880-480EA 203.08 USD
101880-480 101880-482 101880-484
5-Amino-1-(5-chloro-2-pyridinyl)-1H-pyrazole-4-carbonitrile ≥95%
5-Amino-1-(5-chloro-2-pyridinyl)-1H-pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C₉H₆ClN₅
Melting Pt: 272…274 °C
MDL Number: MFCD11841004
CAS Number: 1176567-03-5

Order Now

Specification Test Results

Melting point 272-274°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR