2-Hydrazino-6-methoxybenzothiazole ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Hydrazinyl-6-methoxybenzo[d]thiazole
047514-500MG
047514-5G
101880-400EA
153.85
USD
101880-400
101880-404
2-Hydrazino-6-methoxybenzothiazole ≥95%
2-Hydrazino-6-methoxybenzothiazole
Hazard: Irritant
Formula:
C₈H₉N₃OS MW: 195.241 g/mol Melting Pt: 164-167°C |
MDL Number:
MFCD01439956 CAS Number: 20174-70-3 |
Specification Test Results
Melting point | 164-167°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...