Methyl-2-chloro-6-methoxyisonicotinate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-chloro-6-methoxypyridine-4-carboxylate, Methyl 2-chloro-6-methoxyisonicotinate
047512-1G
047512-5G
101880-392EA
84.62
USD
101880-392
102551-376
Methyl-2-chloro-6-methoxyisonicotinate ≥97%
Methyl 2-chloro-6-methoxyisonicotinate
Hazard: Irritant
Formula:
C₈H₈ClNO₃ MW: 201.61 g/mol Melting Pt: 97.5°C |
MDL Number:
MFCD00173929 CAS Number: 42521-10-8 |
Specification Test Results
Melting point | 97.5°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...