4-[2-(4-Benzylpiperazino)ethoxy]benzenecarboxylicacid ≥95%

Supplier: Matrix Scientific

047510-500MG 047510-5G
101880-378EA 356.92 USD
101880-378 101880-382
4-[2-(4-Benzylpiperazino)ethoxy]benzenecarboxylicacid ≥95%
4-[2-(4-Benzylpiperazino)ethoxy]benzenecarboxylicacid

Hazard: Irritant

Formula: C₂₀H₂₄N₂O₃
MW: 340.43 g/mol
Melting Pt: 185…187 °C
MDL Number: MFCD09027115
CAS Number: 937604-08-5

Order Now

Specification Test Results

Melting point 185-187°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR