2-[4-(2,1,3-Benzothiadiazol-5-ylmethyl)piperazino]-1-ethanol ≥95%

Supplier: Matrix Scientific

Synonyms: 1-[4-(2,1,3-Benzothiadiazol-5-ylmethyl)-4-(2-hydroxyethyl)piperazine

047506-500MG 047506-5G
101880-362EA 556.92 USD
101880-362 101880-366
2-[4-(2,1,3-Benzothiadiazol-5-ylmethyl)piperazino]-1-ethanol ≥95%
2-[4-(2,1,3-Benzothiadiazol-5-ylmethyl)piperazino]-1-ethanol

Hazard: Irritant

Formula: C₁₃H₁₈N₄OS
MW: 278.38 g/mol
Melting Pt: 111…113 °C
MDL Number: MFCD06808731
CAS Number: 874834-85-2

Order Now

Specification Test Results

Melting point 111-113°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR