Methyl-3-{2-[4-(benzyloxy)phenoxy]ethoxy}benzenecarboxylate ≥95%

Supplier: Matrix Scientific

047501-500MG 047501-5G
101880-334EA 278.46 USD
101880-334 101880-338
Methyl-3-{2-[4-(benzyloxy)phenoxy]ethoxy}benzenecarboxylate ≥95%
Methyl-3-{2-[4-(benzyloxy)phenoxy]ethoxy}benzenecarboxylate

Hazard: Irritant

Formula: C₂₃H₂₂O₅
MW: 378.43 g/mol
Melting Pt: 113…115 °C
MDL Number: MFCD08689799
CAS Number: 937602-29-4

Order Now

Specification Test Results

Melting point 113-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR