3-Chloro-N-(3,4-dichlorophenyl)-1-propanesulfonamide ≥95%

Supplier: Matrix Scientific

047499-500MG 047499-5G
101880-322EA 181.54 USD
101880-322 101880-326
3-Chloro-N-(3,4-dichlorophenyl)-1-propanesulfonamide ≥95%
3-Chloro-N-(3,4-dichlorophenyl)-1-propanesulfonamide

Hazard: Irritant

Formula: C₉H₁₀Cl₃NO₂S
Melting Pt: 94…96 °C
MDL Number: MFCD09152740
CAS Number: 952182-56-8

Order Now

Specification Test Results

Melting point 94-96°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR