Ethyl-1-(2,1,3-benzothiadiazol-5-ylmethyl)-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

047495-500MG 047495-5G
101880-298EA 556.92 USD
101880-298 101880-302
Ethyl-1-(2,1,3-benzothiadiazol-5-ylmethyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(2,1,3-benzothiadiazol-5-ylmethyl)-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₉N₃O₂S
MW: 305.4 g/mol
Melting Pt: 48…50 °C
MDL Number: MFCD06808682
CAS Number: 874834-12-5

Order Now

Specification Test Results

Melting point 48-50°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR