Ethyl-1-{3-[4-(benzyloxy)phenoxy]propyl}-3-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

047493-500MG 047493-5G
101880-286EA 375.38 USD
101880-286 101880-290
Ethyl-1-{3-[4-(benzyloxy)phenoxy]propyl}-3-piperidinecarboxylate ≥95%
Ethyl-1-{3-[4-(benzyloxy)phenoxy]propyl}-3-piperidinecarboxylate

Hazard: Irritant

Formula: C₂₄H₃₁NO₄
MW: 397.52 g/mol
Melting Pt: 67…69 °C
MDL Number: MFCD08689797
CAS Number: 937602-25-0

Order Now

Specification Test Results

Melting point 67-69°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR