3-Fluoro-4-(trifluoromethyl)thiobenzamide ≥95%

Supplier: Matrix Scientific

047489-500MG 047489-5G
101880-262EA 267.69 USD
101880-262 101880-266
3-Fluoro-4-(trifluoromethyl)thiobenzamide ≥95%
3-Fluoro-4-(trifluoromethyl)thiobenzamide

Hazard: Irritant

Formula: C₈H₅F₄NS
Melting Pt: 102-104°C
MDL Number: MFCD07369414
CAS Number: 317319-16-7

Order Now

Specification Test Results

Melting point 102-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR