4-Chlorophenacyl thiocyanate ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(4-Chlorophenyl)-2-thiocyanatoethanone, 2-(4-Chlorophenyl)-2-oxoethyl thiocyanate

047487-500MG 047487-5G
101880-250EA 181.54 USD
101880-250 101880-254
4-Chlorophenacyl thiocyanate ≥95%
4-Chlorophenacyl thiocyanate

Hazard: Irritant

Formula: C₉H₆ClNOS
MW: 211.668 g/mol
Melting Pt: 136…138 °C
MDL Number: MFCD00019771
CAS Number: 19339-59-4

Order Now

Specification Test Results

Melting point 136-138°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR