4-Bromophenacyl thiocyanate ≥97%
Supplier: Matrix Scientific
Synonyms:
1-(4-Bromophenyl)-2-thiocyanatoethanone, 2-(4-Bromophenyl)-2-oxoethyl thiocyanate
047485-500MG
047485-5G
101880-238EA
181.54
USD
101880-238
101880-242
4-Bromophenacyl thiocyanate ≥97%
4-Bromophenacyl thiocyanate
Hazard: Irritant
Formula:
C₉H₆BrNOS MW: 256.119 g/mol Melting Pt: 148-149°C |
MDL Number:
MFCD00019770 CAS Number: 65679-14-3 |
Specification Test Results
Melting point | 148-149°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...