Ethyl-4-[4-(tert-butyl)phenyl]-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

047483-500MG 047483-5G
101880-230EA 181.54 USD
101880-230 101880-234
Ethyl-4-[4-(tert-butyl)phenyl]-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-4-[4-(tert-butyl)phenyl]-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₁₉H₂₆N₂O₃
Melting Pt: 160…162 °C
MDL Number: MFCD10543515
CAS Number: 1000927-59-2

Order Now

Specification Test Results

Melting point 160-162°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR