Ethyl-1,6-dimethyl-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

047481-500MG 047481-5G
101880-218EA 181.54 USD
101880-218 101880-222
Ethyl-1,6-dimethyl-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-1,6-dimethyl-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₃
Melting Pt: 175…176 °C
MDL Number: MFCD00205132
CAS Number: 50628-42-7

Order Now

Specification Test Results

Melting point 175-176°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR