Ethyl-4-(3-chlorophenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Supplier: Matrix Scientific
047479-500MG
047479-5G
101880-206EA
181.54
USD
101880-206
101880-210
Ethyl-4-(3-chlorophenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-4-(3-chlorophenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate
Hazard: Irritant
Formula:
C₁₅H₁₇ClN₂O₃ Melting Pt: 135…137 °C |
MDL Number:
MFCD02972684 CAS Number: 512840-35-6 |
Specification Test Results
Melting point | 135-137°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...