1-(4-Bromobenzyl)-4-phenyl-1H-1,2,3-triazol-5-ylamine ≥95%

Supplier: Matrix Scientific

047471-500MG 047471-5G
101880-168EA 556.92 USD
101880-168 101880-172
1-(4-Bromobenzyl)-4-phenyl-1H-1,2,3-triazol-5-ylamine ≥95%
1-(4-Bromobenzyl)-4-phenyl-1H-1,2,3-triazol-5-ylamine

Hazard: Irritant

Formula: C₁₅H₁₃BrN₄
Melting Pt: 152 °C
MDL Number: MFCD06797395
CAS Number: 885950-28-7

Order Now

Specification Test Results

Melting point 152°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR