3-(Hydroxymethyl)-6-methyl-2(1H)-quinolinone ≥95%
Supplier: Matrix Scientific
047463-500MG
047463-5G
101880-120EA
375.38
USD
101880-120
101880-124
3-(Hydroxymethyl)-6-methyl-2(1H)-quinolinone ≥95%
3-(Hydroxymethyl)-6-methyl-2(1H)-quinolinone
Hazard: Irritant
Formula:
C₁₁H₁₁NO₂ Melting Pt: 194 °C (subl.) |
MDL Number:
MFCD02590669 CAS Number: 485337-96-0 |
Specification Test Results
Melting point | 194SUB°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...