3-(4-Chlorobenzyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-ol ≥95%

Supplier: Matrix Scientific

047453-500MG 047453-5G
101880-064EA 556.92 USD
101880-064 101880-068
3-(4-Chlorobenzyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-ol ≥95%
3-(4-Chlorobenzyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-ol

Hazard: Irritant

Formula: C₁₁H₈ClN₅O
MW: 261.67 g/mol
Melting Pt: 183 °C
MDL Number: MFCD07774406
CAS Number: 132269-53-5

Order Now

Specification Test Results

Melting point 183°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR