2-Oxo-1,2-dihydro-3-quinolinecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxyquinoline-3-carboxylic acid, 1,2-Dihydro-2-oxoquinoline-3-carboxylic acid

047447-500MG 047447-5G
101880-028EA 230.77 USD
101880-028 101880-032
2-Oxo-1,2-dihydro-3-quinolinecarboxylic acid ≥95%
2-Oxo-1,2-dihydro-3-quinolinecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
Melting Pt: 330…332 °C
MDL Number: MFCD01550054
CAS Number: 2003-79-4

Order Now

Specification Test Results

Melting point 330-332°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR