2-Chloro-3-quinolinecarbaldehyde oxime ≥95%

Supplier: Matrix Scientific

047440-500MG 047440-5G
101879-992EA 375.38 USD
101879-992 101879-996
2-Chloro-3-quinolinecarbaldehyde oxime ≥95%
2-Chloro-3-quinolinecarbaldehyde oxime

Hazard: Irritant

Formula: C₁₀H₇ClN₂O
Melting Pt: 151…152 °C
MDL Number: MFCD00160598
CAS Number: 93299-49-1

Order Now

Specification Test Results

Melting point 151-152°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR