1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxamide ≥95%
Supplier: Matrix Scientific
047437-500MG
047437-5G
101879-976EA
375.38
USD
101879-976
101879-980
1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxamide ≥95%
1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxamide
Hazard: Irritant
Formula:
C₁₁H₁₁BrN₄O Melting Pt: 188 °C |
MDL Number:
MFCD07780132 CAS Number: 952182-48-8 |
Specification Test Results
Melting point | 188°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...