6-Methyl-2-oxo-1,2-dihydro-3-quinolinecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Methyl-2-oxo-1,2-dihydroquinoline-3-carbaldehyde, 2-Hydroxy-6-methylquinoline-3-carbaldehyde

047434-500MG 047434-5G
101879-960EA 375.38 USD
101879-960 101879-964
6-Methyl-2-oxo-1,2-dihydro-3-quinolinecarbaldehyde ≥95%
6-Methyl-2-oxo-1,2-dihydro-3-quinolinecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₉NO₂
MW: 187.2 g/mol
Melting Pt: >250 °C
MDL Number: MFCD02227050
CAS Number: 101382-53-0

Order Now

Specification Test Results

Melting point >250°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR