3-Nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

047427-500MG 047427-5G
101879-932EA 556.92 USD
101879-932 101879-936
3-Nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine ≥95%
3-Nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₄F₃N₃O₂
Melting Pt: 118…119 °C
MDL Number: MFCD12827812
CAS Number: 1186404-88-5

Order Now

Specification Test Results

Melting point 118-119°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR