7-Fluoro-1-benzothiophene-2-carbohydrazide ≥95%

Supplier: Matrix Scientific

047372-500MG 047372-5G
101879-706EA 375.38 USD
101879-706 101879-710
7-Fluoro-1-benzothiophene-2-carbohydrazide ≥95%
7-Fluoro-1-benzothiophene-2-carbohydrazide

Hazard: Irritant

Formula: C₉H₇FN₂OS
Melting Pt: 231…233 °C
MDL Number: MFCD11100241
CAS Number: 1048913-86-5

Order Now

Specification Test Results

Melting point 231-233°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR