2-Cyano-3-(1H-pyrrol-2-yl)acrylic acid ≥95%

Supplier: Matrix Scientific

047368-500MG 047368-5G
101879-688EA 278.46 USD
101879-688 101879-692
2-Cyano-3-(1H-pyrrol-2-yl)acrylic acid ≥95%
2-Cyano-3-(1H-pyrrol-2-yl)acrylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂
Melting Pt: >300 °C
MDL Number: MFCD11100239
CAS Number: 122520-93-8

Order Now

Specification Test Results

Melting point >300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR